2-Nitrophenylselenocyanate, 98%, Thermo Scientific Chemicals
2-Nitrophenylselenocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitrophenylselenocyanate, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 293580010
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.20 
Save 13.55 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.20 
Save 13.55 (15%)
Add to cart
2-Nitrophenylselenocyanate, 98%, Thermo Scientific Chemicals
Catalog number293580010
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.20 
Save 13.55 (15%)
-
Add to cart
Chemical Identifiers
CAS51694-22-5
IUPAC Name[(2-nitrophenyl)selanyl]formonitrile
Molecular FormulaC7H4N2O2Se
InChI KeyLHBLJWULWKQRON-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1[Se]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Infrared spectrumConforms
Appearance (Form)Powder or crystals or chunks
Melting point139°C to 145°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only