Thermo Scientific Chemicals

Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals

Catalog number: 293610250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals

Catalog number: 293610250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 293610250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2071-20-7
IUPAC Name
[(diphenylphosphanyl)methyl]diphenylphosphane
Molecular Formula
C25H22P2
InChI Key
XGCDBGRZEKYHNV-UHFFFAOYSA-N
SMILES
C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Crystalline powder
Appearance (Color)
White to off-white
Melting point
118°C to 122°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text