Thermo Scientific Chemicals

(R)-(-)-3-Quinuclidinol, 99+%, Thermo Scientific Chemicals

Catalog number: 293730025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-(-)-3-Quinuclidinol, 99+%, Thermo Scientific Chemicals

Catalog number: 293730025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 293730025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
25333-42-0
IUPAC Name
(3R)-1-azabicyclo[2.2.2]octan-3-ol
Molecular Formula
C7H13NO
InChI Key
IVLICPVPXWEGCA-ZETCQYMHSA-N
SMILES
O[C@H]1CN2CCC1CC2
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Specific optical rotation
-43° to -46° (20°C, 589 nm) (c=3, 1 N HCl)
Appearance (Form)
Crystalline powder
Melting point
217°C to 224°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text