Thermo Scientific Chemicals

Tetrakis(trimethylsilyl)silane, 97%, Thermo Scientific Chemicals

Catalog number: 293900050
5 g, Each
Thermo Scientific Chemicals

Tetrakis(trimethylsilyl)silane, 97%, Thermo Scientific Chemicals

Catalog number: 293900050
5 g, Each
Quantity
Catalog number: 293900050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4098-98-0
IUPAC Name
1,1,1,3,3,3-hexamethyl-2,2-bis(trimethylsilyl)trisilane
Molecular Formula
C12H36Si5
InChI Key
BOJSDHZZKKYWAS-UHFFFAOYSA-N
SMILES
C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C
Appearance (Color)
White to beige
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text