4-Bromo-2-fluorophenol, 98%, Thermo Scientific Chemicals
4-Bromo-2-fluorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-2-fluorophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 293921000
Price (USD)/ Each
288.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
288.00
Add to cart
4-Bromo-2-fluorophenol, 98%, Thermo Scientific Chemicals
Catalog number293921000
Price (USD)/ Each
288.00
-
Add to cart
Chemical Identifiers
CAS2105-94-4
IUPAC Name4-bromo-2-fluorophenol
Molecular FormulaC6H4BrFO
InChI KeyRYVOZMPTISNBDB-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to brown
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.5660 to 1.5680 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only