3-Bromo-4-fluorobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Bromo-4-fluorobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-4-fluorobenzaldehyde, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 293930050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS77771-02-9
IUPAC Name3-bromo-4-fluorobenzaldehyde
Molecular FormulaC7H4BrFO
InChI KeyFAHZIKXYYRGSHF-UHFFFAOYSA-N
SMILESFC1=CC=C(C=O)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White to light yellow
Appearance (Form)Low melting solid
Melting point28°C to 33°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only