Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals

Catalog number: 294161000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals

Catalog number: 294161000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 294161000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
489-01-0
IUPAC Name
2,6-di-tert-butyl-4-methoxyphenol
Molecular Formula
C15H24O2
InChI Key
SLUKQUGVTITNSY-UHFFFAOYSA-N
SMILES
COC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Appearance (Form)
Crystalline powder or crystals
Melting point
102°C to 106°C
Appearance (Color)
White to beige
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text