Thermo Scientific Chemicals

1,4,7-Triazacyclononane trihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 294315000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

1,4,7-Triazacyclononane trihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 294315000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 294315000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
58966-93-1
Appearance (Color)
Off-white to yellow
Appearance (Form)
Crystalline powder
Titration Argentometric
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text