Thermo Scientific Chemicals

Cyclen, 97%, Thermo Scientific Chemicals

Catalog number: 294320010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclen, 97%, Thermo Scientific Chemicals

Catalog number: 294320010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 294320010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
294-90-6
IUPAC Name
1,4,7,10-tetraazacyclododecane
Molecular Formula
C8H20N4
InChI Key
QBPPRVHXOZRESW-UHFFFAOYSA-N
SMILES
C1CNCCNCCNCCN1
Appearance (Form)
Crystalline powder
Appearance (Color)
White to light yellow
Melting point
108°C to 113°C
GC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text