Thermo Scientific Chemicals

(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific Chemicals

Catalog number: 294510010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific Chemicals

Catalog number: 294510010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 294510010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29684-56-8
IUPAC Name
(methoxycarbonyl)[(triethylazaniumyl)sulfonyl]azanide
Molecular Formula
C8H18N2O4S
InChI Key
YSHOWEKUVWPFNR-UHFFFAOYSA-N
SMILES
CC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OC
Appearance (Form)
Powder or crystals and/or chunks
Assay
>=96.0 %
Appearance (Color)
White to yellow to light brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text