(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific Chemicals
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 294510050
Price (USD)/ Each
271.65
Online exclusive
302.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
271.65
Online exclusive
302.00 
Save 30.35 (10%)
Add to cart
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific Chemicals
Catalog number294510050
Price (USD)/ Each
271.65
Online exclusive
302.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS29684-56-8
IUPAC Name(methoxycarbonyl)[(triethylazaniumyl)sulfonyl]azanide
Molecular FormulaC8H18N2O4S
InChI KeyYSHOWEKUVWPFNR-UHFFFAOYSA-N
SMILESCC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Assay>=96.0 %
Appearance (Color)White to yellow to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only