Thermo Scientific Chemicals

(1R,2R)-(+)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine, 99%, Thermo Scientific Chemicals

Catalog number: 294662500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

(1R,2R)-(+)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine, 99%, Thermo Scientific Chemicals

Catalog number: 294662500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 294662500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
118628-68-5
IUPAC Name
methyl[(1R,2R)-2-(methylazaniumyl)-1,2-diphenylethyl]azanium
Molecular Formula
C16H22N2
InChI Key
BTHYVQUNQHWNLS-HZPDHXFCSA-P
SMILES
C[NH2+][C@@H]([C@H]([NH2+]C)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Powder or crystals
Melting point
48°C to 51°C
Specific optical rotation
+48° to +51° (20°C, 589 nm) (c=2, MeOH)
Appearance (Color)
White to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text