(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 99%, 99% ee, Thermo Scientific Chemicals
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 99%, 99% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 99%, 99% ee, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 294682500
Price (USD)/ Each
56.00
-
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
56.00
Add to cart
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 99%, 99% ee, Thermo Scientific Chemicals
Catalog number294682500
Price (USD)/ Each
56.00
-
Add to cart
Chemical Identifiers
CAS35132-20-8
IUPAC Name(1R,2R)-1,2-diphenylethane-1,2-bis(aminium)
Molecular FormulaC14H18N2
InChI KeyPONXTPCRRASWKW-ZIAGYGMSSA-P
SMILES[NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=98.0 %
Appearance (Form)Powder or crystalline powder or crystals
Melting point80°C to 84°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only