Thermo Scientific Chemicals

4-Pyridylacetic Acid Hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 294750010
1 g, Each
Thermo Scientific Chemicals

4-Pyridylacetic Acid Hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 294750010
1 g, Each
Quantity
Catalog number: 294750010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6622-91-9
IUPAC Name
2-(pyridin-4-yl)acetic acid hydrochloridyl
Molecular Formula
C7H7ClNO2
InChI Key
NDCASQYPYLKYOP-UHFFFAOYSA-N
SMILES
[Cl].OC(=O)CC1=CC=NC=C1
Appearance (Form)
Crystalline powder or crystals and/or chunks
Titration Argentometric
>=97.5 %
Infrared spectrum
Conforms
Appearance (Color)
White to cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text