Thermo Scientific Chemicals

Samarium, 99.9%, (trace metal basis), 20 to 40 mesh, Thermo Scientific Chemicals

Catalog number: 294780500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Samarium, 99.9%, (trace metal basis), 20 to 40 mesh, Thermo Scientific Chemicals

Catalog number: 294780500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 294780500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-19-9
IUPAC Name
samarium
Molecular Formula
Sm
InChI Key
KZUNJOHGWZRPMI-UHFFFAOYSA-N
SMILES
[Sm]
Appearance (Form)
Powder or filings
Particle size
20 to 40 mesh
Appearance (Color)
Silver grey
Total trace metal impurities
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text