Thermo Scientific Chemicals

Tri-(2-furyl)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: 294800050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Tri-(2-furyl)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: 294800050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 294800050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5518-52-5
IUPAC Name
tris(furan-2-yl)phosphane
Molecular Formula
C12H9O3P
InChI Key
DLQYXUGCCKQSRJ-UHFFFAOYSA-N
SMILES
O1C=CC=C1P(C1=CC=CO1)C1=CC=CO1
Melting point
61°C to 64°C
Appearance (Color)
Off-white to light beige
Appearance (Form)
Crystalline powder
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text