Thermo Scientific Chemicals

4-Quinolinecarboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 294820010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Quinolinecarboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 294820010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 294820010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4363-93-3
IUPAC Name
quinoline-4-carbaldehyde
Molecular Formula
C10H7NO
InChI Key
MGCGJBXTNWUHQE-UHFFFAOYSA-N
SMILES
O=CC1=C2C=CC=CC2=NC=C1
Appearance (Color)
Off-white to yellow to light brown
Infrared spectrum
Conforms
GC
>=94 %
Appearance (Form)
Crystalline powder and/or chunks or solid
Melting point
45°C to 52°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text