Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals
Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals

Sebacoyl chloride, 92%, C10H16Cl2O2, CAS Number-111-19-3, decanedioic acid chloride, sebacoyl chloride, cp, octane-1,8-dicarbonyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, sebacoyl chloride, sebacoylchloride, acmc-1byrk, decanedioyl chloride, 500g, 90% min. (GC)
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
Packaging:
Glass bottle
Catalog number 294900500
Price (USD)/ Each
46.65
Online exclusive
52.10 
Save 5.45 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
46.65
Online exclusive
52.10 
Save 5.45 (10%)
Add to cart
Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals
Catalog number294900500
Price (USD)/ Each
46.65
Online exclusive
52.10 
Save 5.45 (10%)
-
Add to cart
Chemical Identifiers
CAS111-19-3
IUPAC Namedecanedioyl dichloride
Molecular FormulaC10H16Cl2O2
InChI KeyWMPOZLHMGVKUEJ-UHFFFAOYSA-N
SMILESClC(=O)CCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration RCOCl>=90 %
Appearance (Color)Light yellow to golden to brown
AppearanceMay develop some turbidity or precipitate
Refractive index1.4670 to 1.4700 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only