Thermo Scientific Chemicals

Boron tribromide, 99.9%, Thermo Scientific Chemicals

Catalog number: 295205000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Boron tribromide, 99.9%, Thermo Scientific Chemicals

Catalog number: 295205000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 295205000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10294-33-4
IUPAC Name
tribromoborane
Molecular Formula
BBr3
InChI Key
ILAHWRKJUDSMFH-UHFFFAOYSA-N
SMILES
BrB(Br)Br
Appearance (Color)
Clear colorless to amber
Appearance (Form)
Liquid
Assay
>=99.85 % (By difference)
Heavy metals (as Pb)
=<100 ppm
Sulfur (S)
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text