2-Amino-1,3,5-triazine, 98%, Thermo Scientific Chemicals
2-Amino-1,3,5-triazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-1,3,5-triazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 295240050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4122-04-7
IUPAC Name1,3,5-triazin-2-amine
Molecular FormulaC3H4N4
InChI KeyKCZIUKYAJJEIQG-UHFFFAOYSA-N
SMILESNC1=NC=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with HClO4>=97.5 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only