Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number 295530250
Price (USD)/ Each
903.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1063.00 
Save 159.35 (15%)
-
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)/ Each
903.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1063.00 
Save 159.35 (15%)
Add to cart
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Catalog number295530250
Price (USD)/ Each
903.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1063.00 
Save 159.35 (15%)
-
Add to cart
Chemical Identifiers
CAS107-21-1
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
Infrared spectrumConforms
Sulfated ash=<0.005 %
Iron (Fe)=<0.5 ppm
Water=<0.1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only