Thermo Scientific Chemicals

Chloromethyl butyrate, 99%, Thermo Scientific Chemicals

Catalog number: 295580050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Chloromethyl butyrate, 99%, Thermo Scientific Chemicals

Catalog number: 295580050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 295580050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
33657-49-7
IUPAC Name
chloromethyl butanoate
Molecular Formula
C5H9ClO2
InChI Key
BDPZFQLKFUONAG-UHFFFAOYSA-N
SMILES
CCCC(=O)OCCl
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=98.5 %
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4190 to 1.4220 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text