Glycerol, for analysis, 86-88% wt% aqueous solution
Glycerol, for analysis, 86-88% wt% aqueous solution
Glycerol, for analysis, 86-88% wt% aqueous solution
Glycerol, for analysis, 86-88% wt% aqueous solution
Thermo Scientific Chemicals

Glycerol, for analysis, 86-88% wt% aqueous solution

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 295600100
Price (USD)
-
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Heavy metals (as Pb)=<1 ppm
Free acid (CH3COOH)=<0.002 %
Density(20/4°C) 1.224 to 1.232 g/ml
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468051Certificate of AnalysisJan 31, 2025295600025, 295600100, 295602500, 295600010
A0466386Certificate of AnalysisOct 03, 2024295600025, 295600100, 295602500, 295600010
A0462029Certificate of AnalysisApr 18, 2024295600025, 295600100, 295602500, 295600010
A0456941Certificate of AnalysisOct 23, 2023295600025, 295600100, 295602500, 295600010
A0413213Certificate of AnalysisDec 13, 2022295600025, 295600100, 295602500, 295600010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection