Thermo Scientific Chemicals

(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals

Catalog number: 295770250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals

Catalog number: 295770250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 295770250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3724-43-4
IUPAC Name
chloro(dimethylamino)methylium chloride
Molecular Formula
C3H7Cl2N
InChI Key
QQVDYSUDFZZPSU-UHFFFAOYSA-M
SMILES
[Cl-].CN(C)[CH+]Cl
Appearance (Color)
Light yellow to light brown
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Titration Argentometric
95 to 105 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text