Maltitol, 95%
Maltitol, 95%
Maltitol, 95%
Thermo Scientific Chemicals

Maltitol, 95%

CAS: 585-88-6 | C12H24O11 | 344.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number 295800250
Price (USD)
44.65
Special offer
Online exclusive
Ends: 26-Sep-2025
55.30
Save 10.65 (19%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 26-Sep-2025
55.30
Save 10.65 (19%)
Each
Add to cart
Chemical Identifiers
CAS585-88-6
IUPAC Name4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexane-1,2,3,5,6-pentol
Molecular FormulaC12H24O11
InChI KeyVQHSOMBJVWLPSR-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(10 % in water) Clear colorless
Water=<1 %
Specific optical rotationon anhydrous substance
Melting point147°C to 152°C
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link