Thermo Scientific Chemicals

Maltitol, 95%

Catalog number: 295802500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Maltitol, 95%

Catalog number: 295802500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 295802500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
585-88-6
IUPAC Name
4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexane-1,2,3,5,6-pentol
Molecular Formula
C12H24O11
InChI Key
VQHSOMBJVWLPSR-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)CO
Water
=<1 %
Specific optical rotation
on anhydrous substance
Melting point
147°C to 152°C
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text