Thermo Scientific Chemicals

(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride, Thermo Scientific Chemicals

Catalog number: 295810250
25 g, Each
Thermo Scientific Chemicals

(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride, Thermo Scientific Chemicals

Catalog number: 295810250
25 g, Each
Quantity
Catalog number: 295810250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
138124-32-0
IUPAC Name
2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}cyclohexyl)amino]methylidene}cyclohexa-2,4-dien-1-one manganese chloride
Molecular Formula
C36H54ClMnN2O2
InChI Key
YRVXCOIDMFNGIJ-UHFFFAOYNA-M
SMILES
[Cl-].[Mn].CC(C)(C)C1=CC(=CNC2CCCCC2NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C
Appearance (Color)
Dark brown
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals and/or chunks
Elemental Analysis
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text