Thermo Scientific Chemicals

Sodium hydrosulfide hydrate, pure, flakes, Thermo Scientific Chemicals

Catalog number: 296200025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Sodium hydrosulfide hydrate, pure, flakes, Thermo Scientific Chemicals

Catalog number: 296200025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 296200025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207683-19-0
IUPAC Name
sodium sulfanide
Molecular Formula
HNaS
InChI Key
HYHCSLBZRBJJCH-UHFFFAOYSA-M
SMILES
[Na+].[SH-]
Appearance (Color)
Yellow to orange to green to red to brown
Appearance
Color changes in the material may occur
Appearance (Form)
Deliquescent flakes
Titration Iodimetric
67.0 to 73.0 % (NaHS)
Sodium sulfide (Na2S)
=<3.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text