Thermo Scientific Chemicals

Cadmium chloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 296330250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Cadmium chloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 296330250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 296330250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10108-64-2
IUPAC Name
cadmium(2+) dichloride
Molecular Formula
CdCl2
InChI Key
YKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Cd++]
Appearance (Form)
Powder
Appearance (Color)
White
Total trace metal impurities
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text