Thermo Scientific Chemicals

(R)-(-)-Glycidyl nosylate, 98%, Thermo Scientific Chemicals

Catalog number: 296410100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-(-)-Glycidyl nosylate, 98%, Thermo Scientific Chemicals

Catalog number: 296410100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 296410100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
115314-17-5
IUPAC Name
[(2R)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate
Molecular Formula
C9H9NO6S
InChI Key
AIHIHVZYAAMDPM-MRVPVSSYSA-N
SMILES
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@H]1CO1
Appearance (Color)
Light yellow
Specific optical rotation
(20°C, 589 nm) (c=2, CHCl3) < -21.5°
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Crystalline powder or needles

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text