(S)-(+)-Glycidyl nosylate, 98%, Thermo Scientific Chemicals
(S)-(+)-Glycidyl nosylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-Glycidyl nosylate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Packaging:
Glass bottle
Catalog number 296420100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS115314-14-2
IUPAC Name[(2S)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate
Molecular FormulaC9H9NO6S
InChI KeyAIHIHVZYAAMDPM-QMMMGPOBSA-N
SMILES[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
GC>=97.5 %
Appearance (Color)Almost white to yellow-brown
Melting point61°C to 65°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only