Diethoxymethane, 99%, pure, stabilized, Thermo Scientific Chemicals
Diethoxymethane, 99%, pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethoxymethane, 99%, pure, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
Packaging:
Glass bottle
Catalog number 296455000
Price (USD)/ Each
55.65
Online exclusive
61.70 
Save 6.05 (10%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
55.65
Online exclusive
61.70 
Save 6.05 (10%)
Add to cart
Diethoxymethane, 99%, pure, stabilized, Thermo Scientific Chemicals
Catalog number296455000
Price (USD)/ Each
55.65
Online exclusive
61.70 
Save 6.05 (10%)
-
Add to cart
Chemical Identifiers
CAS462-95-3
IUPAC Name(ethoxymethoxy)ethane
Molecular FormulaC5H12O2
InChI KeyKLKFAASOGCDTDT-UHFFFAOYSA-N
SMILESCCOCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Stabilizer50 to 150 ppm (BHT)
Appearance (Form)Liquid
GC>=98.5 %
Peroxides=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only