Thermo Scientific Chemicals

(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%, Thermo Scientific Chemicals

Catalog number: 296470050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%, Thermo Scientific Chemicals

Catalog number: 296470050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 296470050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17553-86-5
IUPAC Name
(7aS)-7a-methyl-2,3,5,6,7,7a-hexahydro-1H-indene-1,5-dione
Molecular Formula
C10H12O2
InChI Key
FNYAZSZTENLTRT-JTQLQIEISA-N
SMILES
C[C@]12CCC(=O)C=C1CCC2=O
Infrared spectrum
Conforms
Specific optical rotation
+330° to +360° (20°C, 589 nm) (c=1, toluene)
Appearance (Form)
Crystalline powder or crystals and/or chunks
Melting point
55°C to 64°C
Appearance (Color)
White to yellow to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text