Chemical Identifiers
CAS81-88-9
IUPAC Name9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC28H31ClN2O3
InChI KeyPYWVYCXTNDRMGF-UHFFFAOYSA-N
SMILES[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC
View more