Thermo Scientific Chemicals

Rhodamine B, 98+%

Catalog number: 296571000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Rhodamine B, 98+%

Catalog number: 296571000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 296571000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81-88-9
IUPAC Name
9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular Formula
C28H31ClN2O3
InChI Key
PYWVYCXTNDRMGF-UHFFFAOYSA-N
SMILES
[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC
Appearance (Color)
Dark green
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
HPLC
>=98.0 %
Moisture
=<2.0 % (105°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text