4-Nitrophenyl isocyanate, 98%, Thermo Scientific Chemicals
4-Nitrophenyl isocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenyl isocyanate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 296700250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-isocyanato-4-nitrobenzene
Molecular FormulaC7H4N2O3
InChI KeyGFNKTLQTQSALEJ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)N=C=O
Molecular Weight (g/mol)164.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Melting point56°C to 62°C
GC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only