Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals
Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 296810025
Price (USD)/ Each
283.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
333.00 
Save 49.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
283.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
333.00 
Save 49.35 (15%)
Add to cart
Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals
Catalog number296810025
Price (USD)/ Each
283.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
333.00 
Save 49.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10217-52-4
IUPAC Namehydrazine
Molecular FormulaH4N2
InChI KeyOAKJQQAXSVQMHS-UHFFFAOYSA-N
SMILESNN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration with HCl35.0 to 35.6 % (N2H4)
Titration with HCl54.5 to 55.5 % (N2H4.xH2O)
Refractive index1.3860 to 1.3900 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only