Thermo Scientific Chemicals

Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals

Catalog number: 296815000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals

Catalog number: 296815000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 296815000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10217-52-4
IUPAC Name
hydrazine
Molecular Formula
H4N2
InChI Key
OAKJQQAXSVQMHS-UHFFFAOYSA-N
SMILES
NN
Appearance (Color)
Clear colorless
Appearance (Form)
Solution
Titration with HCl
35.0 to 35.6 % (N2H4)
Titration with HCl
54.5 to 55.5 % (N2H4.xH2O)
Refractive index
1.3860 to 1.3900 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text