Cyclopropyl Carbinol, 99%, Thermo Scientific Chemicals
Cyclopropyl Carbinol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropyl Carbinol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
100 mL
250 mL
Packaging:
Glass bottle
Catalog number 296820010
Price (USD)/ Each
684.00
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
684.00
Add to cart
Cyclopropyl Carbinol, 99%, Thermo Scientific Chemicals
Catalog number296820010
Price (USD)/ Each
684.00
-
Add to cart
Chemical Identifiers
CAS2516-33-8
IUPAC Namecyclopropylmethanol
Molecular FormulaC4H8O
InChI KeyGUDMZGLFZNLYEY-UHFFFAOYSA-N
SMILESOCC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4290 to 1.4330 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only