Glycine tert butyl ester hydrochloride, 99%
Glycine tert butyl ester hydrochloride, 99%
Thermo Scientific Chemicals

Glycine tert butyl ester hydrochloride, 99%

 
Have Questions?
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number 297020025
Price (USD)/ Each
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-(tert-butoxy)-2-oxoethan-1-aminium
Molecular FormulaC6H14NO2
InChI KeySJMDMGHPMLKLHQ-UHFFFAOYSA-O
SMILESCC(C)(C)OC(=O)C[NH3+]
Molecular Weight (g/mol)132.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration with HClO4>=98.5 %
TLC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only