Thermo Scientific Chemicals

β-Ionone, 96%, synthetic, Thermo Scientific Chemicals

Catalog number: 297131000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

β-Ionone, 96%, synthetic, Thermo Scientific Chemicals

Catalog number: 297131000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 297131000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14901-07-6
IUPAC Name
(3E)-4-(2,6,6-trimethylcyclohex-1-en-1-yl)but-3-en-2-one
Molecular Formula
C13H20O
InChI Key
PSQYTAPXSHCGMF-BQYQJAHWSA-N
SMILES
CC(=O)\C=C\C1=C(C)CCCC1(C)C
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=95 %
Specific gravity
(25°C) 0.941 to 0.947

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text