Thermo Scientific Chemicals

Hydroxypropyl-β-cyclodextrin, 97%, Thermo Scientific Chemicals

Catalog number: 297565000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Hydroxypropyl-β-cyclodextrin, 97%, Thermo Scientific Chemicals

Catalog number: 297565000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 297565000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
128446-35-5
Molecular Formula
C44H75O36
SMILES
OCC-*.[H][C@@]12O[C@H](CO)[C@@H](O[C@@]3([H])O[C@H](CO)[C@@H](O[C@@]4([H])O[C@H](CO)[C@@H](O[C@@]5([H])O[C@H](CO)[C@@H](O[C@@]6([H])O[C@H](CO)[C@@H](O[C@@]7([H])O[C@H](CO)[C@@H](O[C@@]8([H])O[C@H](CO)[C@@H](O1)[C@H](O)[C@H]8O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O
Molecular Weight (g/mol)
1180.05
Appearance (Form)
Powder
HPLC
=<2.0 % Residual beta-cyclodextrin (On dry substance)
NMR
substituents are located mainly at O(2) of the
Water
=<7.5 % (K.F.)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text