Thermo Scientific Chemicals

L(-)-Lactic acid, lithium salt, 99%, pure, Thermo Scientific Chemicals

Catalog number: 297615000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

L(-)-Lactic acid, lithium salt, 99%, pure, Thermo Scientific Chemicals

Catalog number: 297615000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 297615000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
27848-80-2
IUPAC Name
lithium(1+) 2-hydroxypropanoate
Molecular Formula
C3H5LiO3
InChI Key
GKQWYZBANWAFMQ-UHFFFAOYNA-M
SMILES
[Li+].CC(O)C([O-])=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Titration with HClO4
>=98.5 %
Infrared spectrum
Conforms
Specific optical rotation
-13° to -16° (20°C, 589 nm) (c=1, H2O)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text