Thermo Scientific Chemicals

4-(Trifluoromethoxy)aniline, 99%, Thermo Scientific Chemicals

Catalog number: 297640250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Trifluoromethoxy)aniline, 99%, Thermo Scientific Chemicals

Catalog number: 297640250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 297640250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
461-82-5
IUPAC Name
4-(trifluoromethoxy)aniline
Molecular Formula
C7H6F3NO
InChI Key
XUJFOSLZQITUOI-UHFFFAOYSA-N
SMILES
NC1=CC=C(OC(F)(F)F)C=C1
Appearance (Color)
Clear colorless to yellow to light brown
Refractive index
1.4620 to 1.4640 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text