Thermo Scientific Chemicals

4-(Trifluoromethoxy)benzylamine, 97%, Thermo Scientific Chemicals

Catalog number: 297700050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Trifluoromethoxy)benzylamine, 97%, Thermo Scientific Chemicals

Catalog number: 297700050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 297700050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
93919-56-3
IUPAC Name
[4-(trifluoromethoxy)phenyl]methanaminium
Molecular Formula
C8H9F3NO
InChI Key
DBGROTRFYBSUTR-UHFFFAOYSA-O
SMILES
[NH3+]CC1=CC=C(OC(F)(F)F)C=C1
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4510 to 1.4530 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text