2-Thiophenecarbonitrile, 97%, Thermo Scientific Chemicals
2-Thiophenecarbonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Thiophenecarbonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
10 mL
50 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 298052500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1003-31-2
IUPAC Namethiophene-2-carbonitrile
Molecular FormulaC5H3NS
InChI KeyCUPOOAWTRIURFT-UHFFFAOYSA-N
SMILESN#CC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.5620 to 1.5640 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only