trans-2-Decenal, 95%, Thermo Scientific Chemicals
trans-2-Decenal, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-Decenal, 95%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number 298190250
Price (USD)/ Each
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2E)-dec-2-enal
Molecular FormulaC10H18O
InChI KeyMMFCJPPRCYDLLZ-CMDGGOBGSA-N
SMILESCCCCCCC\C=C\C=O
Molecular Weight (g/mol)154.25
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=94 %
Refractive index1.4510 to 1.4600 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only