Thermo Scientific Chemicals

4-Methyl-2-pentenal, 95%, Thermo Scientific Chemicals

Catalog number: 298351000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

4-Methyl-2-pentenal, 95%, Thermo Scientific Chemicals

Catalog number: 298351000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 298351000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5362-56-1
IUPAC Name
(2Z)-4-methylpent-2-enal
Molecular Formula
C6H10O
InChI Key
RIWPMNBTULNXOH-ARJAWSKDSA-N
SMILES
CC(C)\C=C/C=O
Refractive index
1.4360 to 1.4440 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=94 %
Acid value
=<15 mg KOH/g
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text