Thermo Scientific Chemicals

6,7-Dimethoxy-1-phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 298520010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

6,7-Dimethoxy-1-phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 298520010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 298520010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
63768-20-7
IUPAC Name
(1R)-6,7-dimethoxy-1-phenyl-1,2,3,4-tetrahydroisoquinolin-2-ium
Molecular Formula
C17H20NO2
InChI Key
GZGZWZVAJDFXJK-QGZVFWFLSA-O
SMILES
COC1=CC2=C(C=C1OC)[C@H]([NH2+]CC2)C1=CC=CC=C1
Appearance (Form)
Crystalline chunks
Titration with HClO4
>=98.5 %
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text