Thermo Scientific Chemicals

Methanol-d, for NMR, 99.0 atom % D, Thermo Scientific Chemicals

Catalog number: 299130250
25 mL, Glass Bottle, Each
Thermo Scientific Chemicals

Methanol-d, for NMR, 99.0 atom % D, Thermo Scientific Chemicals

Catalog number: 299130250
25 mL, Glass Bottle, Each
Quantity
Packaging
Catalog number: 299130250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1455-13-6
IUPAC Name
methan(²H)ol
Molecular Formula
CH4O
InChI Key
OKKJLVBELUTLKV-VMNATFBRSA-N
SMILES
[2H]OC
Appearance (Form)
Liquid
Water
=<0.1 % (Coulometric)
D-Enrichment
>=99.0 %
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text